missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sec-Butoxide 98.0+%, TCI America™

Catalog No. p-7116785
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A024425G 25 g
A0244500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A024425G Supplier TCI America Supplier No. A024425G
Only null left

Chemical Identifiers

CAS 2269-22-9
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00009327
InChI Key LWGPRERTOLVRLK-UHFFFAOYSA-N
Synonym aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
PubChem CID 50931103
IUPAC Name aluminum;butan-2-ol
SMILES CCC(C)O.CCC(C)O.CCC(C)O.[Al]

Specifications

Color Yellow
Boiling Point 206°C
Quantity 25 g
UN Number 1992
Formula Weight 246.33
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Aluminum sec-Butoxide
EINECSNumber (2)-2388
TSCA Yes