missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sec-butoxide, 97%

Catalog No. AAA1304422
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304422 100 g
AAA1304436 500 g
AAA130440E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1304422 Supplier Thermo Scientific Chemicals Supplier No. A1304422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2269-22-9 | C12H30AlO3 | 249.351 g/mol

Aluminum sec-butoxide is used to prepare photosensitive alumina gel films. It is also used in the preparation of microporous crystalline aluminophosphate AlPO-41 and alumina mesoporous material. It is used as a gelling agent of paint and ink and water repellent substance of wood. Further, it acts as an active catalyst and reducer of aldehydes and ketones. In addition to this, it is used to make high-surface-area alumina using 2,4-pentanedione as a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum sec-butoxide is used to prepare photosensitive alumina gel films. It is also used in the preparation of microporous crystalline aluminophosphate AlPO-41 and alumina mesoporous material. It is used as a gelling agent of paint and ink and water repellent substance of wood. Further, it acts as an active catalyst and reducer of aldehydes and ketones. In addition to this, it is used to make high-surface-area alumina using 2,4-pentanedione as a chelating agent.

Solubility
Miscible with alcohol, isopropyl alcohol and toluene.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, metals and halogens.

Chemical Identifiers

CAS 2269-22-9
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00009327
InChI Key LWGPRERTOLVRLK-UHFFFAOYSA-N
Synonym aluminum sec-butoxide, aluminum tri-sec-butoxide, 2-butanol, aluminum salt, aluminum sec-butylate, aluminum butan-2-olate, aluminum tri-s-butoxide, 2-butanol, aluminum salt 3:1, aluminum tri-sec-butanolate, al o-sec-bu 3, aluminiumtri-sec-butoxide
PubChem CID 50931103
IUPAC Name aluminum;butan-2-ol
SMILES CCC(C)O.CCC(C)O.CCC(C)O.[Al]

Specifications

Boiling Point 200°C to 206°C (30mmHg)
Quantity 100 g
UN Number UN1992
Beilstein 3910908
Flash Point 27°C (82°F)
Solubility Information Miscible with alcohol,isopropyl alcohol and toluene.
Formula Weight 246.32
Percent Purity 97%
Odor Alcohol-like
Refractive Index 1.439
Sensitivity Moisture sensitive
Density 0.967
Chemical Name or Material Aluminum sec-butoxide
Show More Show Less
Hazard Category H226-H319-H335-H336
Hazard Statement GHS H Statement
H226-H302-H312-H315-H319
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 218-871-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only