missing translation for 'onlineSavingsMsg'
Learn More

Aluminum rod, 6.35mm (0.25in) dia, Puratronic™, 99.999% (metals basis)

Catalog No. p-7048794
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 cm
4 x 25 cm
5 cm
Catalog No. Quantity
AA11493BQ 5 cm
AA11493BU 25 cm
AA11493CZ 4 x 25 cm
3 options

Catalog No. AA11493BQ

Supplier: Thermo Scientific Chemicals 011493BQ

Only null left

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 5 cm
Odor Odorless
Weight ≈0.87g/cm
Form Rod
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum rod, 6.35mm (0.25 in.) dia.
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only