missing translation for 'onlineSavingsMsg'
Learn More

Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9995% (metals basis)

Catalog No. AA43429BU
Change view
Click to view available options
Quantity:
5 cm
25 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43429BU 25 cm
AA43429BQ 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43429BU Supplier Thermo Scientific Chemicals Supplier No. 043429BU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7429-90-5 | Al | 26.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Quantity 25 cm
Odor Odorless
Weight ≈10.35g/cm
Form Rod
Percent Purity 99.9995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Aluminum rod, 22mm (0.87 in.) dia.
EINECSNumber 231-072-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only