missing translation for 'onlineSavingsMsg'
Learn More

Aluminum powder, spherical, APS 3.0-4.5 micron, 97.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4408685
Change view
Click to view available options
Quantity:
1 kg
5 kg
Catalog No. Quantity
AA41000A1 1 kg
AA41000A7 5 kg
2 options

Catalog No. AA41000A1

Supplier: Thermo Scientific Chemicals 041000A1

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Moisture Sensitive; Tap density 1.4g/cm3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver to Gray
Appearance Silver Gray powder
Quantity 1 kg
Odor Odorless
Sensitivity Water reactive
Form Powder
Assay Percent Range 97.5% (Metals basis)
Chemical Name or Material Aluminum, APS 3.0-4.5 micron
Hazard Category H250-H261-H500
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 231-072-3
RTECSNumber BD0330000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only