missing translation for 'onlineSavingsMsg'
Learn More

Aluminum powder, spherical, APS 17-30 micron, 99% (metals basis), Thermo Scientific Chemicals

$406.78 - $406.78

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA10576A3
View Documents
Thermo Scientific Chemicals
010576A3
2 kg N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
aluminium
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629
[Al]

Specifications

Silver to Gray
Silver Gray powder
400°C (752°F)
Water reactive
99% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium
5359268
Aluminum
7429-90-5
2 kg
Odorless
Powder
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Videos
Safety and Handling

Safety and Handling

P210-P222-P223-P231+P232-P280-P335+P334-P370+P378s-P501c

H250-H261-H500

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 231-072-3

RTECSNumber : BD0330000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only