missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Technical, Spectrum™ Chemical

Catalog No. 18608545
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608545 500 g Poly Bottle
18608546 2.5 kg Poly Bottle
18608547 12 kg Poly Bottle
18608548 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608545 Supplier Spectrum Chemical Mfg Cor Supplier No. A1106500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7784-24-9

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Chemical Name or Material Aluminum Potassium Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.