missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate Dodecahydrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6002326
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002326 500 g
6002325 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002326 Supplier Solstice Supplier No. 31242500G

Puriss. p.a., ACS Reagent, Reagent European Pharmacopoeia,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Quantity 500 g
Linear Formula AlK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 474.39g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density 1.757g/mL at 25°C
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate, Reag. Ph. Eur.
EINECSNumber 233-141-3
RTECSNumber WS5690000