missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate dodecahydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 99.0-100.5% (calc. on dry substance), Solstice

Catalog No. 60044740
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044740 6 x 1 kg
6001944 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044740 Supplier Solstice Supplier No. 126236X1KG

Puriss., meets analytical specification of BP, European Pharmacopoeia, 99.0-100.5% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
MDL Number MFCD00149143
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Melting Point 92°C (literature)
CAS Max % 100.0000%
Quantity 6 x 1 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula AlK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 474.39
Percent Purity 99.0 to 100.5% (calc. on dry substance)
Grade puriss.
Packaging Poly Bottle
Density 1.757 g/mL at 25 °C(lit.)
Chemical Name or Material Aluminum potassium sulfate dodecahydrate, Puriss., Meets analytical specification of BP, Ph. Eur., 99.0-100.5% (calc. on dry substance)
Show More Show Less
ShelfLife 1650 days from date of manufacture
EINECSNumber 233-141-3
RTECSNumber WS5690000
Recommended Storage Room Temp