missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate Dodecahydrate, Purified (Potassium Alum), Reagents

Catalog No. 01337509
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337509 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337509 Supplier Labchem Inc Supplier No. C2127500500A5

Aluminum Potassium Sulfate Dodecahydrate, Purified (Potassium Alum), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.37
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym Alum
IUPAC Name aluminium(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Boiling Point 200°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.725
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate, Purified (Potassium Alum)