missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Crystal, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. p-7156910
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18600484 500 g Poly Bottle
18600485 2.5 kg Poly Bottle
18600486 12 kg Poly Pail
18600487 50 kg Fiber Drum
4 options

Catalog No. 18600484

Supplier: Spectrum Chemical Mfg Cor A1107500GM

Only null left

A1107, 7784-24-9, AlK(SO4)2·12H2O

Spectrum™ Chemical Aluminum Potassium Sulfate, Crystal, FCC is commonly used as an additive to baking powder to provide a second leavening phase at high temperatures. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.37
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
IUPAC Name aluminum(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade FCC
Packaging Poly Bottle