missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Potassium Sulfate, Crystal, FCC, 99.5-100.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical Aluminum Potassium Sulfate, Crystal, FCC is commonly used as an additive to baking powder to provide a second leavening phase at high temperatures. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 2.5 kg |
| Percent Purity | 99.5 to 100.5% |
| Grade | FCC |
| Packaging | Poly Bottle |