missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Crystal, FCC, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18600484
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600484 500 g Poly Bottle
18600485 2.5 kg Poly Bottle
18600486 12 kg Poly Bottle
18600487 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600484 Supplier Spectrum Chemical Mfg Cor Supplier No. A1107500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1107, 7784-24-9, AlK(SO4)2·12H2O

Spectrum™ Chemical Aluminum Potassium Sulfate, Crystal, FCC is commonly used as an additive to baking powder to provide a second leavening phase at high temperatures. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.37
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
IUPAC Name aluminum(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity 99.5 to 100.5%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Aluminum Potassium Sulfate, Crystal