missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7164966
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18608540 125 g Poly Bottle
18608541 500 g Poly Bottle
18608542 2.5 kg Poly Bottle
18608543 12 kg Poly Pail
18608544 50 kg Fiber Drum
5 options

Catalog No. 18608540

Supplier: Spectrum Chemical Mfg Cor A1105125GM

Only null left

Chemical Identifiers

CAS 7784-24-9

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.