missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18608540
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608540 125 g Poly Bottle
18608541 500 g Poly Bottle
18608542 2.5 kg Poly Bottle
18608543 12 kg Poly Bottle
18608544 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608540 Supplier Spectrum Chemical Mfg Cor Supplier No. A1105125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7784-24-9

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 125 g
Percent Purity 98 to 102%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Aluminum Potassium Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.