missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Potassium Sulfate, 12-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390674
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002253 500 g Poly Bottle
02002252 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002253 Supplier Avantor Supplier No. 054601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.372
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
Synonym kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
PubChem CID 62667
ChEBI CHEBI:86465
IUPAC Name aluminum;potassium;disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 474.39
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Aluminum Potassium Sulfate, 12-Hydrate