missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, plasma standard solution, Specpure™ Al 10,000μg/mL

Catalog No. AA38721AP
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38721AP 500 mL
AA38721AD 50 mL
AA38721AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38721AP Supplier Thermo Scientific Chemicals Supplier No. 038721AP

Used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry. | CAS: 11145-27-0 | AlSi | 55.067 g/mol

Aluminum, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11145-27-0
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00134029
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Physical Form Liquid
Quantity 500 mL
UN Number UN3264
Solubility Information Miscible with water.
Packaging Plastic bottle
Chemical Name or Material Aluminum
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only