missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, Pellets, Spectrum™ Chemical

$696.50 - $12226.03

Chemical Identifiers

CAS 7429-90-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608562
View Documents
Spectrum Chemical Mfg Cor
A1058500GM
500 g Poly Pail
Each for $696.50
Only null left
 
18608561
View Documents
Spectrum Chemical Mfg Cor
A105825KG
2.5 kg Poly Pail
Each for $2,640.53
Only null left
 
18608560
View Documents
Spectrum Chemical Mfg Cor
A105812KGBL
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7429-90-5

Specifications

7429-90-5
500 g
Pellet
Aluminum
1
Poly Pail
Ungraded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.