missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide substrate, 10x10x0.5mm, polished two sides, A plane

Catalog No. AA41426KT
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41426KT 1 Ea.
AA41426KU 5 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41426KT Supplier Thermo Scientific Chemicals Supplier No. 041426KT

Al2O3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Aluminum Oxide Substrate, A Plane
Name Note Polished Two Sides
Quantity 1 Ea.
Molecular Formula Al2O3
MDL Number MFCD00003424

RUO – Research Use Only