missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide Standard, MilliporeSigma™ Supelco™

Catalog No. p-7214098
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022407 1 kg
111022409 5 kg
111022408 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111022407 Supplier Merck Emd Millipore Supplier No. 063001KG

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration. Aluminum oxides are generally used as solid support phases in solid phase extraction (SPE)-based clean-up procedures employed in chromatographic methods.

Specifications

Form Powder
Grade Standard
Quantity 1 kg