missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide Standard, MilliporeSigma™ Supelco™

Catalog No. 111022407
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022407 1 kg
111022409 5 kg
111022408 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111022407 Supplier Merck Emd Millipore Supplier No. 063001KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration. Aluminum oxides are generally used as solid support phases in solid phase extraction (SPE)-based clean-up procedures employed in chromatographic methods.

Specifications

Form Powder
Grade Standard
Quantity 1 kg