missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, single crystal

Catalog No. AA40259KG
Change view
Click to view available options
Quantity:
1 Pc.
5 Pc.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40259KG 1 Pc.
AA40259KJ 5 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40259KG Supplier Thermo Scientific Chemicals Supplier No. 040259KG

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 1 Pc.
IUPAC Name dialuminium(3+) trioxidandiide
Packaging Shrink-wrapped
Chemical Name or Material Aluminum oxide
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only