missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide-silicon oxide (13%), catalyst support, low surface area, macroporous

Catalog No. AA43863A7
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43863A7 5 kg
AA43863A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43863A7 Supplier Thermo Scientific Chemicals Supplier No. 043863A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: S.A. 0.008m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide-silicon oxide is a fibrous material used as loose wool, blanket, felt, paper or boards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Spheres
Quantity 5 kg
Solubility Information Insoluble in water.
Odor Odorless
Appearance White spheres
Chemical Name or Material Aluminum oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only