missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, gamma-phase, nanopowder, 99+%

Catalog No. p-4781772
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AA4475714 25 g
AA4475722 100 g
AA4475736 500 g
3 options

Catalog No. AA4475714

Supplier: Thermo Scientific Chemicals 04475714

Only null left

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is used in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. It can act as a catalyst in a variety of reactions such as the Claus process. It is used in the production of aluminum, manufacture of abrasives, refractories, ceramics, electrical insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >99%
Solubility Information Insoluble in water.
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Odor Odorless
Chemical Name or Material Aluminum oxide
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only