missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, Gamma phase, Catalyst support, High surface area, Bimodal

Catalog No. p-6827162
Change view
Click to view available options
Quantity:
50 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4383218 50 g
AA43832A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4383218 Supplier Thermo Scientific Chemicals Supplier No. 04383218
Only null left

High surface area, Bimodal | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide finds applications in catalyst support, chemical sensors, as an absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Physical Form Pellets
Quantity 50 g
Solubility Information Insoluble in water. Slightly soluble in acid and alkali.
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Density 3.965
Chemical Name or Material Aluminum Oxide, Catalyst Support
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only