Learn More
Aluminum oxide, gamma-phase, alpha-phase, 99.98% (metals basis)
Description
Aluminum oxide finds use as catalyst in organic synthesis and chromatography. It is used as an ingredient in sunscreen lotions and paints. It is also used in the manufacture of abrasives, refractories, ceramics, filler for plastics, dosimeter, spark plug insulators, and electrical insulators. Aluminum oxide is used widely to produce aluminum metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 2980°C |
| Melting Point | 2045°C |
| Physical Form | Powder |
| Quantity | 25 g |
| Merck Index | 14,356 |
| Solubility Information | Insoluble in water,ethanol and ether |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| Formula Weight | 101.96 |
| Percent Purity | 99.98% |
| Odor | Odorless |
| Show More |
Safety and Handling
| EINECSNumber | 215-691-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only