missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, fused, insulating powder, 99.7+%

Used in Dry Filling For thermal Insulation of Electrical Furnaces | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

$158.88 - $513.70

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33200A1
View Documents
Thermo Scientific Chemicals
033200A1
1 kg
Each for $158.88
Only null left
 
AA33200A7
View Documents
Thermo Scientific Chemicals
033200A7
5 kg
Each for $513.70
Only null left
 
Description

Description

Used in dry filling for thermal insulation of electrical furnaces.Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Form: approx. 60 micron
Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)

Specifications

1344-28-1
>99.7%
MFCD00003424
Insoluble in water,but slightly soluble in acid and alkali.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
Odorless
1.33 hPa (1mmHg) at 2158°C
1 kg
Al2O3
aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
White powder
Aluminum oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-691-6

RTECSNumber : BD1200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only