missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, For the Determination of Hydrocarbons, MilliporeSigma™ Supelco™

Catalog No. 111012984
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012984 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012984 Supplier Merck Emd Millipore Supplier No. 05184100GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym Alumina
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point 2040°C
Physical Form Powder
Quantity 100 g
Linear Formula Al2O3
Formula Weight 101.96
RTECSNumber BD1200000