missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, catalyst support, low surface area

Catalog No. p-7076459
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Catalog No. Quantity
AA4386230 250 g
AA43862A1 1 kg
AA43862A7 5 kg
3 options

Catalog No. AA4386230

Supplier: Thermo Scientific Chemicals 04386230

Only null left

Low surface area | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Physical Form Pellets
Quantity 250 g
IUPAC Name dialuminum;oxygen(2-)
Chemical Name or Material Aluminum Oxide, Catalyst Support
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only