missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, catalyst support, low silica

Catalog No. p-4864958
Change view
Click to view available options
Quantity:
1 kg
5 kg
Catalog No. Quantity
AA44849A1 1 kg
AA44849A7 5 kg
2 options

Catalog No. AA44849A1

Supplier: Thermo Scientific Chemicals 044849A1

Only null left

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Spheres
Quantity 1 kg
Appearance White spheres
Chemical Name or Material Aluminum Oxide, Low silica
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only