missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, catalyst support, intermediate surface area (low SiO{2})

Catalog No. p-4864956
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43857A1 1 kg
AA43857A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43857A1 Supplier Thermo Scientific Chemicals Supplier No. 043857A1
Only null left

Form: S.A. 100m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Pellets
Quantity 1 kg
Solubility Information Slightly soluble in acid and alkali. Insoluble in water.
IUPAC Name dialuminium(3+) trioxidandiide
Odor Odorless
Appearance Off-white pellets
Chemical Name or Material Aluminum oxide, Contains 0.1% SiO2
EINECSNumber 215-691-6
RTECSNumber BD1200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only