missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, calcined, insulating powder, Thermo Scientific Chemicals

Catalog No. AA33195A1
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33195A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA33195A1 Supplier Thermo Scientific Chemicals Supplier No. 033195A1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Quantity 1 kg
IUPAC Name dialuminum;oxygen(2-)
Odor Odorless
Chemical Name or Material Aluminum oxide, Calcined, Insulating powder
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only