missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, basic, for TLC

Catalog No. p-7080035
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4600122 100 g
AA4600136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4600122 Supplier Thermo Scientific Chemicals Supplier No. 04600122
Only null left

For TLC | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color Pink to White
Physical Form Powder
Quantity 100 g
Linear Formula Al2O3
Merck Index 14,356
Solubility Information Insoluble in water.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide, Basic
Show More Show Less
EINECSNumber 215-691-6
RTECSNumber BD1200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only