Learn More
Aluminum oxide, Alpha-phase, 99.9% (metals basis)
S.A. 6-8m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
$82.26 - $716.71
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4257222
|
Thermo Scientific Chemicals
04257222 |
100 g |
Each for $82.26
|
|
|||||
|
AA4257236
|
Thermo Scientific Chemicals
04257236 |
500 g |
Each for $238.32
|
|
|||||
|
AA42572A3
|
Thermo Scientific Chemicals
042572A3 |
2 kg |
Each for $716.71
|
|
|||||
Description
Aluminum oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
| 2980°C | |
| White | |
| Powder | |
| 99.9% (Metal basis) | |
| MFCD00003424 | |
| Slowly soluble in aqueous alkali solutions-forming hydroxides. Insoluble in water and in non-polar organic solvents. | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| 101.96 | |
| 1.765 | |
| Aluminum oxide |
| 2045°C | |
| 1344-28-1 | |
| 100 g | |
| Al2O3 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminum;oxygen(2-) | |
| 9989226 | |
| Odorless | |
| White powder |
Safety and Handling
EINECSNumber : 215-691-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only