Learn More
Aluminum oxide, Aerosol Refractory Paint, Thermo Scientific Chemicals
$443.70 - $1724.00
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| IUPAC Name | dialuminium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA40391CLC
|
Thermo Scientific Chemicals
040391CLC |
13 oz. |
Each for $443.70
|
|
|||||
|
AA403910CL
|
Thermo Scientific Chemicals
0403910CL |
5 x 13 oz. |
Pack for $1,724.00
|
|
|||||
Description
Provides easy application of uniform, thin, protective high temperature Aluminum oxide coatings. Substrates can be ceramic, graphite or metal. Aluminum oxide is also widely used as an abrasive. In the paper-making process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Provides easy application of uniform, thin, protective high temperature Al2O3 coatings
- Substrates can be ceramic, graphite or metal
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| dialuminium(3+) trioxidandiide |
Specifications
| -44°C | |
| 1344-28-1 | |
| Al2O3 | |
| UN1950 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| 9989226 | |
| Aluminum oxide, Aerosol Refractory Paint |
| White | |
| 13 oz. | |
| MFCD00003424 | |
| -104°C (-155°F) | |
| Slightly soluble in water. insoluble in diethyl ether Practically insoluble in ethanol | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| Aerosol Spray Can |
Safety and Handling
GHS H Statement
H222-H319-H336
Extremely flammable aerosol.
Causes serious eye irritation.
May cause drowsiness or dizziness.
P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
H222-H319-H336-H350-X
DOTInformation : Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
TSCA : Yes
Recommended Storage : Ambient temperatures
For Manufacturing and Laboratory Use. Not for beverage use