Learn More
Aluminum oxide, Aerosol Refractory Brushable Paint
Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
$201.16 - $738.86
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
Provides protective high temperature Al{2}O{3} coating on ceramic, graphite, or metalAluminum oxide, Aerosol Refractory Brushable Paint is used as barrier layer between dissimilar materials to prevent reaction and interactions during high temperature. It also acts as a solvent-based paint. It also provides protective high temperature alumina coating on ceramic, graphite and metal.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
| White | |
| Paste | |
| Al2O3 | |
| UN1263 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminium(3+) trioxidandiide | |
| 9989226 |
| 1344-28-1 | |
| 250 g | |
| MFCD00003424 | |
| −4°C (25°F) | |
| Slightly soluble in water. | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| Aluminum oxide, Aerosol Refractory Brushable Paint |
Safety and Handling
GHS H Statement
H225-H319
Highly flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PAINT
TSCA : Yes
For Manufacturing and Laboratory Use. Not for beverage use