missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, Aerosol Refractory Brushable Paint

Catalog No. AA4311930
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4311930 250 g
AA43119A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4311930 Supplier Thermo Scientific Chemicals Supplier No. 04311930

Provides protective high temperature Al2O3 coating on ceramic, graphite, or metal | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Provides protective high temperature Al{2}O{3} coating on ceramic, graphite, or metalAluminum oxide, Aerosol Refractory Brushable Paint is used as barrier layer between dissimilar materials to prevent reaction and interactions during high temperature. It also acts as a solvent-based paint. It also provides protective high temperature alumina coating on ceramic, graphite and metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color White
Physical Form Paste
Quantity 250 g
UN Number UN1263
Flash Point −4°C (25°F)
Solubility Information Slightly soluble in water.
IUPAC Name dialuminium(3+) trioxidandiide
Chemical Name or Material Aluminum oxide, Aerosol Refractory Brushable Paint
Hazard Category H336
Hazard Statement GHS H Statement
H225-H319
Highly flammable liquid and vapor.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PAINT
TSCA Yes

For Manufacturing and Laboratory Use. Not for beverage use