missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Activated, neutral, Brockmann I, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| pH | Neutral |
| Quantity | 1 kg |
| Linear Formula | Al2O3 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 101.96g/mol |
| Chemical Name or Material | Aluminum Oxide, Activated Brockmann I |
Safety and Handling
| EINECSNumber | 215-691-6 |
| RTECSNumber | BD1200000 |