missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Activated Brockmann I, Honeywell Fluka™

Catalog No. 60045083
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045083 1 kg Poly Bottle
60045084 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045083 Supplier Honeywell Chemicals Supplier No. 1999661KG
Only null left

Activated, acidic, Brockmann I

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2072°C
pH Acidic
Quantity 1 kg
Linear Formula Al2O3
UN Number NONH for all modes of transport
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96g/mol
Packaging Poly Bottle
Appearance Powder
Density 3.960g/cu.cm
Chemical Name or Material Aluminum Oxide, Activated Brockmann I
Show More Show Less
EINECSNumber 215-691-6
RTECSNumber BD1200000