missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide, Activated, basic, Brockmann I, Honeywell Fluka™
Activated, basic, Brockmann I
$388.90 - $2168.38
Chemical Identifiers
| CAS | 1344-28-1 |
|---|---|
| Molecular Formula | Al2O3 |
| Molecular Weight (g/mol) | 101.96 |
| MDL Number | MFCD00003424 |
| InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
| Synonym | aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
| PubChem CID | 9989226 |
| IUPAC Name | dialuminum;oxygen(2-) |
| SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 1344-28-1 | |
| 101.96 | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| 9989226 | |
| [O--].[O--].[O--].[Al+3].[Al+3] |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| dialuminum;oxygen(2-) |
Specifications
| 1344-28-1 | |
| 1 kg | |
| Al2O3 | |
| NONH for all modes of transport | |
| PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
| dialuminum;oxygen(2-) | |
| 9989226 | |
| Aluminum Oxide, Activated Brockmann I |
| Basic | |
| Al2O3 | |
| MFCD00003424 | |
| aluminum oxide, aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
| [O--].[O--].[O--].[Al+3].[Al+3] | |
| 101.96 | |
| 101.96g/mol |
Safety and Handling
EINECSNumber : 215-691-6
RTECSNumber : BD1200000