missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Acid Technical A, EcoChrom™, MP Biomedicals™

Catalog No. ICN07895
Change view
Click to view available options
Quantity:
50 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN07895 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN07895 Supplier MP Biomedicals Inc Supplier No. 0207895

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Alumina is essentially aluminum oxide, Al2O3, occurring in nature as minerals bauxite, bayerite, corundum, etc.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Color Pink to White
Physical Form Free-flowing Powder
Quantity 50 kg
Formula Weight 101.96
Density 3.94g/cm3 at 20°C
Recommended Storage Store at room temperature.