missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, 99% (metals basis)

Catalog No. AA1255336
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1255336 500 g
AA12553A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1255336 Supplier Thermo Scientific Chemicals Supplier No. 01255336

Catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

It finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Physical Form Fine Powder
Quantity 500 g
Assay Percent Range 99% (metals basis)
Solubility Information Aluminum oxide is insoluble in water,but slightly soluble in acid and alkali.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Refractive Index 1.768
Density 3.965 g/mL
Chemical Name or Material Aluminum oxide
Show More Show Less
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only