missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide 60 G neutral, EMD-Millipore

Supplier:  Technidata 1010902500

Catalog No. M1010902500

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Aluminum oxide 60 G neutral
2.5 kg
aluminum oxides-PLC, aluminum oxides-TLC
Density: 4.0g/cm3 (20°C); Bulk Density: 500 kg/m3

Chemical Identifiers

aluminum oxides-PLC, aluminum oxides-TLC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store without restrictions

SDS
Documents

Documents

Promotions

Promotions