missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, 5016-A, Basic, Brockmann Grade I

Catalog No. AA11503A7
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11503A7 5 kg
AA1150330 250 g
AA11503A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA11503A7 Supplier Thermo Scientific Chemicals Supplier No. 011503A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol

Aluminum oxide is used as an abrasive, dosimeter, filler of choice for plastics, ingredient in sunscreen and cosmetics. Aluminum oxide flakes are also used in paint for providing reflective decorative effects. In laboratories it is widely used as a medium for chromatographic purifications. It is used in the production of aluminum (Hall–Héroult process), alumina nanofibers, cutoff tools, and sandpaper. It serves as a catalyst for several reactions of industrial importance, including converting alcohols into alkenes, decomposing hydrazine, drying of gases, and converting hydrogen sulfide into sulfur in refineries (Claus Process). It is also used in integrated circuits, compact fluorescent lamps (CFL), sodium vapor lamps, hip replacements, CD/DVD polishing, superconducting quantum interference devices (SQUIDS), insulation devices, and as a support for many popular catalysts (for example, in Ziegler-Natta polymerizations).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point 2045°C
Color White
Physical Form Powder
Quantity 5 kg
Solubility Information Insoluble in water,diethyl ether and ethanol.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Brockmann
Odor Odorless
Chemical Name or Material Aluminum oxide
EINECSNumber 215-691-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only