Learn More
Eisco™ Aluminum Overflow Can - 4 Inch
Eisco Labs aluminum overflow can is ideal for liquid displacement experiments, providing accurate and reliable results in laboratory and educational settings.
Supplier:  Eisco™ PH119BSINGLE
Catalog No. S02903
Only null left
Description
Description
  • 4 inch tall, 2 inch diameter
  • Used for liquid displacement experiments
  • With Spout
Specifications
Specifications
Overflow Can
Aluminum
Product Suggestions
Product Suggestions