missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitride Rod, Round; Diameter (mm), 10

Catalog No. AA42998DR
Change view
Click to view available options
Quantity:
200 mm
300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42998DR 200 mm
AA42998DW 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42998DR Supplier Thermo Scientific Chemicals Supplier No. 42998DR

CAS: 24304-00-5 | AlN | 40.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24304-00-5
Molecular Formula AlN
Molecular Weight (g/mol) 40.99
MDL Number MFCD00003429
InChI Key PIGFYZPCRLYGLF-UHFFFAOYSA-N
Synonym aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c
PubChem CID 90455
ChEBI CHEBI:50884
IUPAC Name alumanylidyneamine
SMILES N#[Al]

Specifications

Color White to Yellow
Melting Point >2200°C
Appearance White to pale yellow rod
Density 3.26
Quantity 200 mm
Odor Odorless
Form Rod
Chemical Name or Material Aluminum nitride
EINECSNumber 246-140-8
RTECSNumber BD1055000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only