missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

Supplier:  Thermo Scientific Chemicals 04314522

 View more versions of this product

Catalog No. AA4314522


Only null left
Explore available promotions

Description

Description

Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum nitrate nonahydrate
Low mercury
Crystalline
(metals basis)
MFCD00003419
14,352
Soluble in water (540mg/ml),ethanol,alcohol (very),and acetone (slightly).
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
375.129
375.13 (212.99 Anhydrous)
Puratronic
Hygroscopic
73.5°C (135° (decomposition))
7784-27-2
100 g
AlH18N3O18
UN1438
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
SWCIQHXIXUMHKA-UHFFFAOYSA-N
aluminum;trinitrate;nonahydrate
24567
99.999%
Odorless

Chemical Identifiers

7784-27-2
375.129
SWCIQHXIXUMHKA-UHFFFAOYSA-N
24567
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]
AlH18N3O18
MFCD00003419
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
aluminum;trinitrate;nonahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.

P280-P305+P351+P338-P310

H318

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: ALUMINUM NITRATE

EINECSNumber : 236-751-8

RTECSNumber : BD1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only