missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis)

Catalog No. AA4314514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4314514 25 g
AA4314522 100 g
AA4314536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4314514 Supplier Thermo Scientific Chemicals Supplier No. 04314514

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00003419
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.5°C (135° (decomposition))
Physical Form Crystalline
Quantity 25 g
UN Number UN1438
Merck Index 14,352
Solubility Information Soluble in water (540mg/ml),ethanol,alcohol (very),and acetone (slightly).
Formula Weight 375.13 (212.99 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Aluminum nitrate nonahydrate, Low mercury
Show More Show Less
Hazard Category H318
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: ALUMINUM NITRATE
EINECSNumber 236-751-8
RTECSNumber BD1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only