missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg)

Catalog No. p-4407192
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
500 g
Catalog No. Quantity
AA1062606 5 g
AA1062614 25 g
AA1062622 100 g
AA1062636 500 g
4 options

Catalog No. AA1062606

Supplier: Thermo Scientific Chemicals 01062606

Only null left

CAS: 25838-59-9 | AlN3O9 | 212.99 g/mol

Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25838-59-9
Molecular Formula AlN3O9
Molecular Weight (g/mol) 212.99
MDL Number MFCD00003419
InChI Key JLDSOYXADOWAKB-UHFFFAOYSA-N
Synonym aluminum nitrate hydrate, al.3no3.h2o, aluminum nitrate hydrate, puratronic, aluminum 3+ hydrate trinitrate
PubChem CID 22078336
SMILES [Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 73.5°C
Color White
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.999% (Metals basis excluding Hg)
Solubility Information Soluble in water,methanol,ethanol,and ethylene glycol.
IUPAC Name aluminium(3+) trinitrate
Formula Weight 212.99 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 236-751-8
RTECSNumber BD1050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only