missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Metal, 200 Mesh, Powder, Spectrum™ Chemical

$353.90 - $12243.47

Chemical Identifiers

CAS 7429-90-5
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18608520
View Documents
Spectrum Chemical Mfg Cor
A105025GM
25 g Poly Bottle
Each for $353.90
Only null left
 
18608524
View Documents
Spectrum Chemical Mfg Cor
A1050500GM
500 g Poly Bottle
Each for $935.40
Only null left
 
18608522
View Documents
Spectrum Chemical Mfg Cor
A1050100LBBL
100 lb. Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18608523
View Documents
Spectrum Chemical Mfg Cor
A105025KG
2.5 kg Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18608521
View Documents
Spectrum Chemical Mfg Cor
A105012KG
12 kg Poly Bottle N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7429-90-5

Specifications

7429-90-5
25 g
Powder
Aluminum Metal
1
Poly Bottle
Ungraded
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.